Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3-cyclohexene-1-carboxylate | 1049004-32-1 | 280.17 | 5 G
SDP

Catalog No. 5000446179
Encompass_Preferred

Bpin-Cyclohexene-COOEt, also known as Ethyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3-cyclohexene-1-carboxylate, is a PROTAC linker belonging to the PEG class. It is designed for use in the synthesis of PROTAC molecules. This compound is a colorless to light yellow liquid.

  • Intended for research use only.
  • Unstable in solutions, requiring freshly prepared solutions for optimal use.
  • Molecular weight of 280.17.
  • Purity of 97.0%.

Catalog No. 50-004-46179 Supplier Medchemexpress LLC Supplier No. HYW0066355G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bpin-Cyclohexene-COOEt, also known as Ethyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3-cyclohexene-1-carboxylate, is a PROTAC linker belonging to the PEG class. It is designed for use in the synthesis of PROTAC molecules. This compound is a colorless to light yellow liquid.

  • Intended for research use only.
  • Unstable in solutions, requiring freshly prepared solutions for optimal use.
  • Molecular weight of 280.17.
  • Purity of 97.0%.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.