Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 4,4,4-Trifluoro-3-hydroxybutyrate 98.0+%, TCI America™
SDP

Catalog No. T19495G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T19495G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T19495G Supplier TCI America Supplier No. T19495G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 372-30-5
Molecular Formula C6H9F3O3
Molecular Weight (g/mol) 186.13
MDL Number MFCD00040842
InChI Key ZWEDFBKLJILTMC-UHFFFAOYNA-N
Synonym ethyl 3-hydroxy-4,4,4-trifluorobutyrate, ethyl 4,4,4-trifluoro-3-hydroxybutyrate, ethyl 3-hydroxy-4,4,4-trifluorobutanoate, 3-hydroxy-4,4,4-trifluorobutyric acid ethyl ester, 4,4,4-trifluoro-3-hydroxybutyric acid ethyl ester, ethyl3-hydroxy-4,4,4-trifluorobutyrate, butanoic acid, 4,4,4-trifluoro-3-hydroxy-, ethyl ester, acmc-209irs, acmc-20al28, ksc496k8b
PubChem CID 136210
IUPAC Name ethyl 4,4,4-trifluoro-3-hydroxybutanoate
SMILES CCOC(=O)CC(O)C(F)(F)F

Specifications

Color White
Boiling Point 83°C
Quantity 5 g
Formula Weight 186.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Ethyl 4,4,4-Trifluoro-3-hydroxybutyrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.