Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethyl 4,4,4-trifluoroacetoacetate

Catalog No. E5020525G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E5020525G 25 g
E50205100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E5020525G Supplier Sigma Aldrich Supplier No. E5020525G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Ethyl 4,4,4-trifluoroacetoacetate (etfaa) is a general reagent to synthesize enantiopure trifluoromethyl-functionalized products. applications include:
  • Synthesis of (s)- and (r)-α-trifluoromethyl-aspartic acid and α- trifluoromethyl-serine from chiral cf3-oxazolidines, which is derived from etfaa
  • Enantiopure synthesis of trifluoromethyl-α-amino acid derivatives
  • Synthesis of (2r)-2-trifluoromethyl-2-carboxyazetidine, (r)- and (s)-trifluoromethylhomoserines from oxazolidine intermediate obtained by condensing (r)-phenylglycinol with etfaa

Specifications

CAS 372-31-6
Density 1.259 g/mL (at 25°C (literature))
Boiling Point 129°C to 130°C (lit.)
Molecular Formula C6H7F3O3
Refractive Index n20/D 1.375 (literature)
Linear Formula CF3COCH2CO2C2H5
MDL Number MFCD00000424
Quantity 25 g
Synonym ETFAA; Ethyl 3-oxo-4,4,4-trifluorobutyrate
Molecular Weight (g/mol) 184.11
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.