Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 4,4,4-trifluoroacetoacetate, 97%, Thermo Scientific Chemicals

Catalog Number AC119030250
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
372-31-6
C6H7F3O3
184.11
MFCD00000424
OCJKUQIPRNZDTK-UHFFFAOYSA-N
ethyl 4,4,4-trifluoroacetoacetate, ethyl trifluoroacetoacetate, butanoic acid, 4,4,4-trifluoro-3-oxo-, ethyl ester, 4,4,4-trifluoro-3-oxobutanoic acid ethyl ester, ethyl trifluoroacetyl acetate, etfaa, 4,4,4-trifluoroacetoacetic acid ethyl ester, ethyl-4,4,4-trifluoroacetoacetate, trifluoroacetoacetic acid ethyl ester, 4,4,4-trifluoro-3-oxo-butyric acid ethyl ester
67793
ethyl 4,4,4-trifluoro-3-oxobutanoate
CCOC(=O)CC(=O)C(F)(F)F
372-31-6
C6H7F3O3
184.11
MFCD00000424
OCJKUQIPRNZDTK-UHFFFAOYSA-N
ethyl 4,4,4-trifluoroacetoacetate, ethyl trifluoroacetoacetate, butanoic acid, 4,4,4-trifluoro-3-oxo-, ethyl ester, 4,4,4-trifluoro-3-oxobutanoic acid ethyl ester, ethyl trifluoroacetyl acetate, etfaa, 4,4,4-trifluoroacetoacetic acid ethyl ester, ethyl-4,4,4-trifluoroacetoacetate, trifluoroacetoacetic acid ethyl ester, 4,4,4-trifluoro-3-oxo-butyric acid ethyl ester
67793
ethyl 4,4,4-trifluoro-3-oxobutanoate
CCOC(=O)CC(=O)C(F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -39.0°C
Color Colorless to Yellow
Density 1.2500g/mL
Boiling Point 131.0°C (709.0 mmHg)
Flash Point 28°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.3750 to 1.3780
Linear Formula CF3COCH2CO2C2H5
Quantity 25 mL
Beilstein 03, II, 425
Specific Gravity 1.25
Solubility Information Solubility in water: 10g/L (20°C). Other solubilities: miscible with ethanol and ether
Formula Weight 184.11
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Ethyl 4, 4, 4-trifluoroacetoacetate, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Flammable liquid and vapor.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid release to the environment.
EINECSNumber 206-750-7

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit