Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 4,4,4-trifluorobutyrate, 98%, Thermo Scientific™

Catalog No. AC309650050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC309650050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC309650050 Supplier Thermo Scientific Chemicals Supplier No. 309650050

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 371-26-6
Molecular Formula C6H9F3O2
Molecular Weight (g/mol) 170.13
MDL Number MFCD00041398
InChI Key PSRZMXNNQTWAGB-UHFFFAOYSA-N
Synonym ethyl 4,4,4-trifluorobutyrate, ethyl4,4,4-trifluorobutyrate, 4,4,4-trifluorobutyric acid ethyl ester, 4,4,4-trifluoro-butyric acid ethyl ester, butanoic acid, 4,4,4-trifluoro-, ethyl ester, acmc-1cncd, ethyl-4,4,4-trifluorobutyrate, ethyl 3-trifluoromethylpropionate, butanoic acid,4,4,4-trifluoro-, ethyl ester
PubChem CID 2733273
IUPAC Name ethyl 4,4,4-trifluorobutanoate
SMILES CCOC(=O)CCC(F)(F)F

Specifications

Color Colorless to Yellow
Density 1.1600g/mL
Boiling Point 127.0°C
Flash Point 33°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.3510 to 1.3530
Linear Formula CF3CH2CH2CO2C2H5
Quantity 5 g
Specific Gravity 1.16
Formula Weight 170.13
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl 4,4,4-trifluorobutyrate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapour.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Keep away fr

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.