Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 4,4,5,5,5-Pentafluoro-3-oxovalerate 96.0+%, TCI America™
SDP

Catalog No. P10625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P10625G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P10625G Supplier TCI America Supplier No. P10625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 663-35-4
Molecular Formula C7H7F5O3
Molecular Weight (g/mol) 234.12
MDL Number MFCD00013569
InChI Key MWGSZQXKIYWSFS-UHFFFAOYSA-N
Synonym ethyl pentafluoropropionylacetate, ethyl 4,4,5,5,5-pentafluoro-3-oxovalerate, ethyl pentafluoropropionyl acetate, ethyl pentafluoropropanoyl acetate, 4,4,5,5,5-pentafluoro-3-oxovaleric acid ethyl, pentanoic acid, 4,4,5,5,5-pentafluoro-3-oxo-, ethyl ester, acmc-209nuz, ethyl pentafluoropropionyl ace tate
PubChem CID 522203
IUPAC Name ethyl 4,4,5,5,5-pentafluoro-3-oxopentanoate
SMILES CCOC(=O)CC(=O)C(F)(F)C(F)(F)F

Specifications

Color Yellow
Boiling Point 64°C
Quantity 5 g
UN Number 3272
Formula Weight 234.12
Percent Purity ≥96.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Ethyl 4,4,5,5,5-Pentafluoro-3-oxovalerate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.