Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 4,4-Difluoroacetoacetate 96.0+%, TCI America™
SDP

Catalog No. E101825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E101825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E101825G Supplier TCI America Supplier No. E101825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 352-24-9
Melting Point -46°C
Color Yellow
Boiling Point 160°C
Molecular Formula C6H8F2O3
Quantity 25 g
MDL Number MFCD00039277
Synonym ethyl 4,4-difluoroacetoacetate, ethyl 4,4-difluoro-3-oxobutyrate, ethyl difluoroacetoacetate, butanoic acid, 4,4-difluoro-3-oxo-, ethyl ester, ethyl4,4-difluoro-3-oxobutyrate, ethyl 4,4-difluoro-3-oxo-butanoate, 4,4-difluoroacetoacetic acid ethyl ester, 4,4-difluoro-3-oxo-butyric acid ethyl ester, ethyl4,4-difluoro-3-oxobutanoate, pubchem21933
InChI Key CBDPWKVOPADMJC-UHFFFAOYSA-N
SMILES CCOC(=O)CC(=O)C(F)F
IUPAC Name ethyl 4,4-difluoro-3-oxobutanoate
Molecular Weight (g/mol) 166.124
PubChem CID 67697
Formula Weight 166.12
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 4,4-Difluoroacetoacetate
Show More Show Less
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.