Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 4-[(4-Methoxybenzylidene)amino]cinnamate 98.0+%, TCI America™
SDP

Catalog No. M06021G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M06021G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M06021G Supplier TCI America Supplier No. M06021G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6421-30-3
Molecular Formula C19H19NO3
Molecular Weight (g/mol) 309.365
MDL Number MFCD00059231
InChI Key FMNNRRPDXZWUAE-KGCZTCBMSA-N
Synonym 4-(p-Anisalamino)cinnamic Acid Ethyl Ester, 4-[(p-Anisylidene)amino]cinnamic Acid Ethyl Ester, Ethyl 4-(p-Anisalamino)cinnamate, Ethyl 4-[(p-Anisylidene)amino]cinnamate, 4-[(4-Methoxybenzylidene)amino]cinnamic Acid Ethyl Ester
PubChem CID 780892
IUPAC Name ethyl (E)-3-[4-[(4-methoxyphenyl)methylideneamino]phenyl]prop-2-enoate
SMILES CCOC(=O)C=CC1=CC=C(C=C1)N=CC2=CC=C(C=C2)OC

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 309.37
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 4-[(4-Methoxybenzylidene)amino]cinnamate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.