Learn More
Medchemexpress LLC Ethyl 4,6-dichloronicotinate | 40296-46-6 | 220.05 | 10 G
SDP
Catalog No. 5000447668
Encompass_Preferred
Ethyl 4,6-dichloronicotinate is a biochemical reagent used in life science research. It functions as a biological material or organic compound and serves as an important raw material and intermediate in various chemical synthesis processes.
  • Used as a biological material or organic compound
  • Applicable for life science related research
  • Important raw material in organic synthesis
  • Intermediate in pharmaceuticals
  • Intermediate in agrochemicals
  • Intermediate in dyestuff
  • Functions as a fine chemical intermediate
Catalog No. 50-004-47668 Supplier Medchemexpress LLC Supplier No. HYW00255810G
Ethyl 4,6-dichloronicotinate is a biochemical reagent used in life science research. It functions as a biological material or organic compound and serves as an important raw material and intermediate in various chemical synthesis processes.
  • Used as a biological material or organic compound
  • Applicable for life science related research
  • Important raw material in organic synthesis
  • Intermediate in pharmaceuticals
  • Intermediate in agrochemicals
  • Intermediate in dyestuff
  • Functions as a fine chemical intermediate