Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 4-Acetyl-5-oxohexanoate 96.0+%, TCI America™
SDP

Catalog No. A15265G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15265G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A15265G Supplier TCI America Supplier No. A15265G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 2832-10-2
Color Yellow
Boiling Point 182°C
Molecular Formula C10H16O4
Quantity 5 g
MDL Number MFCD00009211
Synonym ethyl 4,4-diacetylbutyrate, 4-acetyl-5-oxohexanoic acid ethyl ester, 4,4-diacetylbutyric acid ethyl ester, ethyl 4-acetyl-5-oxohexanoate, 4-acetyl-5-oxo-hexanoic acid ethyl ester, hexanoic acid,4-acetyl-5-oxo-, ethyl ester, hexanoic acid, 4-acetyl-5-oxo-, ethyl ester
InChI Key CVPCFJRJUCNUJG-VQHVLOKHSA-N
SMILES CCOC(=O)CCC(C(=O)C)C(=O)C
IUPAC Name ethyl 4-acetyl-5-oxohexanoate
Molecular Weight (g/mol) 200.23
PubChem CID 2733934
Formula Weight 200.23
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 4-Acetyl-5-oxohexanoate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.