Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 4-acetylbenzoate, 98%, Thermo Scientific™

Catalog No. AAH6325803
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6325803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6325803 Supplier Thermo Scientific Chemicals Supplier No. H6325803

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38430-55-6
Molecular Formula C11H12O3
Molecular Weight (g/mol) 192.21
MDL Number MFCD00013241
InChI Key GLOAPLPTWAXAIG-UHFFFAOYSA-N
Synonym 4-acetylbenzoic acid ethyl ester, benzoic acid, 4-acetyl-, ethyl ester, 4-acetyl-benzoic acid ethyl ester, ethyl4-acetylbenzoate, ethyl 4-acetylbenzate, pubchem18162, ethyl 4-acetyl benzoate, ethyl 4-acetyl-benzoate, acmc-1csm7, ethyl 4-acetylbenzoate
PubChem CID 600911
IUPAC Name ethyl 4-acetylbenzoate
SMILES CCOC(=O)C1=CC=C(C=C1)C(C)=O

Specifications

Melting Point 55°C to 57°C
Quantity 1 g
Formula Weight 192.21
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Ethyl 4-acetylbenzoate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.