Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethyl 4-aminobutanoate hydrochloride | 6937-16-2 | 98.0% | 167.63 | 1 ML
SDP

Catalog No. 5000366926
Encompass_Preferred

Ethyl 4-aminobutanoate hydrochloride is an amino acid derivative that serves as a cornerstone in organic synthesis for creating organic compounds with specific structures and functions. It can also be used to synthesize matrix metalloproteinase (MMP) inhibitors.

  • Amino acid derivative
  • Used in organic synthesis
  • Can be used to synthesize matrix metalloproteinase (MMP) inhibitors
  • For research use only
  • Molecular weight: 167.63
  • Formula: C6H14ClNO2
  • Appearance: solid
  • Color: white to off-white

Catalog No. 50-003-66926 Supplier Medchemexpress LLC Supplier No. HYW01007810MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethyl 4-aminobutanoate hydrochloride is an amino acid derivative that serves as a cornerstone in organic synthesis for creating organic compounds with specific structures and functions. It can also be used to synthesize matrix metalloproteinase (MMP) inhibitors.

  • Amino acid derivative
  • Used in organic synthesis
  • Can be used to synthesize matrix metalloproteinase (MMP) inhibitors
  • For research use only
  • Molecular weight: 167.63
  • Formula: C6H14ClNO2
  • Appearance: solid
  • Color: white to off-white

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.