Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 4-bromo-3-ethoxy-2-butenoate, tech. 90%, stab. with sodium carbonate

Catalog No. AAL0684903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0684903 1 g
AAL0684906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0684903 Supplier Thermo Scientific Chemicals Supplier No. L0684903
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 1116-50-3 | C8H13BrO3 | 237.09 g/mol

Ethyl 4-bromo-3-ethoxy-2-butenoate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 4-bromo-3-ethoxy-2-butenoate is used as pharmaceutical intermediate.

Solubility
Not miscible in water.

Notes
Moisture and light sensitive. Incompatible with oxidizing agents, bases and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1116-50-3
Molecular Formula C8H13BrO3
Molecular Weight (g/mol) 237.09
MDL Number MFCD00051757
InChI Key FYYMWILFRUUBCX-UHFFFAOYSA-N
Synonym ethyl 4-bromo-3-ethoxybut-2-enoate, ethyl 4-bromo-3-ethoxy-2-butenoate, ethyl 2e-4-bromo-3-ethoxybut-2-enoate, ethyl e-4-bromo-3-ethoxybut-2-enoate
PubChem CID 6376123
SMILES CCOC(=O)C=C(CBr)OCC

Specifications

Boiling Point 88°C to 90°C (1 mmHg)
Odor Pungent
Refractive Index 1.496
Quantity 1 g
UN Number UN3265
Beilstein 2042896
Sensitivity Light and moisture sensitive
Solubility Information Not miscible in water.
IUPAC Name ethyl 4-bromo-3-ethoxybut-2-enoate
Formula Weight 237.1
Percent Purity 90%
Grade Technical
Chemical Name or Material Ethyl 4-bromo-3-ethoxy-2-butenoate, stabilized with sodium carbonate
Show More Show Less
Hazard Category H302-H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.