Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethyl 4-bromobutyrate

Catalog No. 167118250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-711-8250G 250 g
16-711-850G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 16-711-8250G Supplier Sigma Aldrich Supplier No. 167118250G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Ethyl 4-bromobutyrate was used in enzyme-linked immunosorbent assay for zilpaterol
  • It was used in the synthesis of isosteric thieno[2,3-b]-azepin-4-ones, having anti-tumor activity
  • It was also used in the synthesis of poly(2,6-dimethyl-1,4-phenylene oxide)

Specifications

CAS 2969-81-5
Density 1.363 g/mL (at 25°C (literature))
Boiling Point 80°C to 82°C (10 mmHg)
Molecular Formula C6H11BrO2
Linear Formula Br(CH2)3 COOC2H5
Refractive Index n20/D 1.456 (literature)
MDL Number MFCD00000259
Quantity 250 g
Molecular Weight (g/mol) 195.05
Percent Purity 95%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.