Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethyl 4-(butylamino) benzoate | 94-32-6 | 100.0% | 221.30 | 25 G
SDP

Catalog No. 5000403175
Encompass_Preferred

Ethyl 4-(butylamino)benzoate is a chemical compound provided as a white to off-white solid. It has a molecular formula of C13H19NO2 and a molecular weight of 221.30. This compound is stable under recommended storage conditions and is intended for laboratory research use only. It has not been fully validated for medical applications.

  • White to off-white solid appearance
  • Melting-freezing point of 70.2-70.4°C
  • Boiling point of 338.4 °C at 760 mmHg
  • Purity of 99.97% (HPLC)
  • Stable under recommended storage conditions
  • For research use only

Catalog No. 50-004-03175 Supplier Medchemexpress LLC Supplier No. 25GHY41462
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethyl 4-(butylamino)benzoate is a chemical compound provided as a white to off-white solid. It has a molecular formula of C13H19NO2 and a molecular weight of 221.30. This compound is stable under recommended storage conditions and is intended for laboratory research use only. It has not been fully validated for medical applications.

  • White to off-white solid appearance
  • Melting-freezing point of 70.2-70.4°C
  • Boiling point of 338.4 °C at 760 mmHg
  • Purity of 99.97% (HPLC)
  • Stable under recommended storage conditions
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.