Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethyl [4-chloro-3-(trifluoromethyl)phenyl]carbamate | 18585-06-3 | MFCD00043450 | 98.0% | 267.63 g·mol-1 | C10H9ClF3NO2 | 250 MG
SDP

Catalog No. 5000209445
Encompass_Preferred

Sorafenib impurity 2 is an impurity standard derived from the kinase inhibitor sorafenib, supplied for analytical and research use. It has CAS No. 18585-06-3, molecular formula C10H9ClF3NO2, and a molecular weight of 267.63 g·mol-1. The compound is provided in small laboratory quantities with reported high purity and should be stored and handled according to safety data guidance.

  • Used as an analytical reference or impurity standard
  • CAS number 18585-06-3 for unambiguous identification
  • Molecular weight 267.63 g·mol-1
  • Available in small laboratory quantities for research
  • Store in solvent at -80 °C for long-term stability
  • Reported purity 98.0%

Catalog No. 50-002-09445 Supplier Medchemexpress LLC Supplier No. 250MGHYW048413
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sorafenib impurity 2 is an impurity standard derived from the kinase inhibitor sorafenib, supplied for analytical and research use. It has CAS No. 18585-06-3, molecular formula C10H9ClF3NO2, and a molecular weight of 267.63 g·mol-1. The compound is provided in small laboratory quantities with reported high purity and should be stored and handled according to safety data guidance.

  • Used as an analytical reference or impurity standard
  • CAS number 18585-06-3 for unambiguous identification
  • Molecular weight 267.63 g·mol-1
  • Available in small laboratory quantities for research
  • Store in solvent at -80 °C for long-term stability
  • Reported purity 98.0%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.