Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethyl 4-chloro-4-oxobutyrate

Catalog No. 190705100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-070-5100G 100 g
19-070-525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-070-5100G Supplier Sigma Aldrich Supplier No. 190705100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Ethyl 4-chloro-4-oxobutyrate was used in the synthesis of:
  • Polymer-supported bifunctional catalyst
  • Succinimidyl ester of benzo(a)pyrene
  • Succinate prodrugs of curcuminoids

Specifications

CAS 14794-31-1
Density 1.155 g/mL (at 25°C (literature))
Boiling Point 88°C to 90°C (11 mmHg)
Molecular Formula C6H9ClO3
Refractive Index n20/D 1.437 (literature)
Linear Formula ClCOCH2CH2CO2C2H5
MDL Number MFCD00000751
Quantity 100 g
Synonym Ethyl succinyl chloride
Molecular Weight (g/mol) 164.59
Percent Purity 95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.