Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 4-chloro-5,7-difluoroquinoline-3-carboxylate, 99%, Thermo Scientific™

Catalog No. AAH50514MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH50514MD 250 mg
AAH5051403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH50514MD Supplier Thermo Scientific Chemicals Supplier No. H50514MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 311346-69-7
Molecular Formula C12H8ClF2NO2
Molecular Weight (g/mol) 271.648
MDL Number MFCD00173416
InChI Key IEGYWXRVOVIAMV-UHFFFAOYSA-N
Synonym ethyl 4-chloro-5,7-difluoro-3-quinolinecarboxylate, 4-chloro-5,7-difluorolquinoline-3-carboxylic acid ethyl ester
PubChem CID 3743835
IUPAC Name ethyl 4-chloro-5,7-difluoroquinoline-3-carboxylate
SMILES CCOC(=O)C1=C(C2=C(C=C(C=C2N=C1)F)F)Cl

Specifications

Quantity 250 mg
Formula Weight 271.65
Percent Purity 99%
Chemical Name or Material Ethyl 4-chloro-5,7-difluoroquinoline-3-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.