Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 4-Chlorobutyrate 98.0+%, TCI America™
SDP

Catalog No. C083025ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C083025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C083025ML Supplier TCI America Supplier No. C083025ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3153-36-4
Molecular Formula C6H11ClO2
Molecular Weight (g/mol) 150.602
MDL Number MFCD00001004
InChI Key OPXNFHAILOHHFO-UHFFFAOYSA-N
Synonym ethyl 4-chlorobutyrate, butanoic acid, 4-chloro-, ethyl ester, ethyl gamma-chlorobutyrate, ethyl-4-chlorobutyrate, 4-chloro-butyric acid ethyl ester, butyric acid, 4-chloro-, ethyl ester, ethyl .gamma.-chlorobutyrate, 4-chlorobutyric acid ethyl ester, ethyl 4chlorobutyrate, acmc-1cqjw
PubChem CID 18474
IUPAC Name ethyl 4-chlorobutanoate
SMILES CCOC(=O)CCCCl

Specifications

Color Yellow
Boiling Point 186°C
Quantity 25 mL
Formula Weight 150.60
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 4-Chlorobutyrate
EINECSNumber (2)-2649&(9)-0213
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.