Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 4-Fluorocinnamate 98.0+%, TCI America™
SDP

Catalog No. E10991G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E10991G 1 g
E10995G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E10991G Supplier TCI America Supplier No. E10991G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 352-03-4
Molecular Formula C11H11FO2
Molecular Weight (g/mol) 194.205
MDL Number MFCD02252626
InChI Key UNAXNPTZJKKUGO-VMPITWQZSA-N
Synonym 4-Fluorocinnamic Acid Ethyl Ester
PubChem CID 5373059
IUPAC Name ethyl (E)-3-(4-fluorophenyl)prop-2-enoate
SMILES CCOC(=O)C=CC1=CC=C(C=C1)F

Specifications

Melting Point 34°C
Color White-Yellow
Boiling Point 131°C
Quantity 1 g
Formula Weight 194.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 4-Fluorocinnamate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.