Learn More
Medchemexpress LLC Ethyl 4-hydroxybenzoate-d4 | 1219795-53-5 | 170.20 | C9H6D4O3 | 10 MG
SDP
Catalog No. 5000296744
Encompass_Preferred
Ethylparaben-d4 is the deuterium-labeled analog of ethyl 4-hydroxybenzoate supplied for research use as an isotopically labeled internal standard or tracer for quantitative analysis by NMR, GC-MS, and LC-MS. It contains four deuterium atoms (d4) to enable mass discrimination and accurate quantitation.
  • Deuterium-labeled internal standard for quantitative analysis.
  • Suitable for NMR, GC-MS, and LC-MS applications.
  • Molecular formula C9H6D4O3 and molecular weight 170.20.
  • Available in small analytical quantities (commonly 1 mg, 5 mg, 10 mg).
  • For research use only; not for human or clinical use.
Catalog No. 50-002-96744 Supplier Medchemexpress LLC Supplier No. HYB0934S10MG
Ethylparaben-d4 is the deuterium-labeled analog of ethyl 4-hydroxybenzoate supplied for research use as an isotopically labeled internal standard or tracer for quantitative analysis by NMR, GC-MS, and LC-MS. It contains four deuterium atoms (d4) to enable mass discrimination and accurate quantitation.
  • Deuterium-labeled internal standard for quantitative analysis.
  • Suitable for NMR, GC-MS, and LC-MS applications.
  • Molecular formula C9H6D4O3 and molecular weight 170.20.
  • Available in small analytical quantities (commonly 1 mg, 5 mg, 10 mg).
  • For research use only; not for human or clinical use.