Learn More
Medchemexpress LLC Ethyl (4-hydroxyphenyl)acetate | 17138-28-2 | MFCD00016491 | 99.3% | 180.20 g·mol⁻¹ | C10H12O3 | 25 G
SDP
Catalog No. 5000303095
Encompass_Preferred
Ethyl (4-hydroxyphenyl)acetate is an organic ester used as a chemical carbon source and reagent in biochemical research and synthetic chemistry. It is a colorless to light yellow compound (C10H12O3, MW 180.20 g·mol⁻¹) supplied in laboratory pack sizes and intended for research applications.
  • Serves as a chemical carbon source for microbial or enzymatic studies.
  • High purity suitable for analytical and preparative work.
  • Available in multiple pack sizes for laboratory use.
  • Colorless to light yellow; physical state varies near room temperature.
  • Suitable as a building block in organic synthesis.
Catalog No. 50-003-03095 Supplier Medchemexpress LLC Supplier No. 25GHYW016483
Ethyl (4-hydroxyphenyl)acetate is an organic ester used as a chemical carbon source and reagent in biochemical research and synthetic chemistry. It is a colorless to light yellow compound (C10H12O3, MW 180.20 g·mol⁻¹) supplied in laboratory pack sizes and intended for research applications.
  • Serves as a chemical carbon source for microbial or enzymatic studies.
  • High purity suitable for analytical and preparative work.
  • Available in multiple pack sizes for laboratory use.
  • Colorless to light yellow; physical state varies near room temperature.
  • Suitable as a building block in organic synthesis.