Learn More
Medchemexpress LLC Ethyl (4-hydroxyphenyl)acetate | 17138-28-2 | MFCD00016491 | 99.3% | 180.20 g/mol | C10H12O3 | 50 G
SDP
Catalog No. 5000301796
Encompass_Preferred
Ethyl (4-hydroxyphenyl)acetate is an ethyl ester used as a chemical carbon source in research and biochemical assays. It is a colorless to light-yellow solid or liquid depending on temperature, has formula C10H12O3 and molecular weight 180.20 g/mol, and is supplied for research use only.
  • High purity (99.28%) suitable for analytical and research use.
  • Useful as a carbon source and reagent in biochemical assays.
  • Available in multiple pack sizes to support small- and large-scale work.
  • Physical state varies with temperature, informing storage and handling.
  • Supplied for research use only; not for human or clinical use.
Catalog No. 50-003-01796 Supplier Medchemexpress LLC Supplier No. HYW01648350G
Ethyl (4-hydroxyphenyl)acetate is an ethyl ester used as a chemical carbon source in research and biochemical assays. It is a colorless to light-yellow solid or liquid depending on temperature, has formula C10H12O3 and molecular weight 180.20 g/mol, and is supplied for research use only.
  • High purity (99.28%) suitable for analytical and research use.
  • Useful as a carbon source and reagent in biochemical assays.
  • Available in multiple pack sizes to support small- and large-scale work.
  • Physical state varies with temperature, informing storage and handling.
  • Supplied for research use only; not for human or clinical use.