Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethyl 4-hydroxyphenylacetate | 17138-28-2 | MFCD00016491 | 99.3% | 180.20 | C10H12O3 | 100 G
SDP

Supplier:  Medchemexpress LLC HYW016483100G

Encompass_Preferred

Ethyl (4-hydroxyphenyl)acetate is an organic ester (C10H12O3, MW 180.20) used as a chemical carbon source and reagent in research applications. It is colorless to light yellow and may be solid or liquid near ambient temperatures; supplied at high purity for laboratory use.

  • Chemical formula: C10H12O3.
  • Molecular weight: 180.20 g/mol.
  • High purity suitable for research (99.28%).
  • Appearance: colorless to light yellow; solid below ~36°C, liquid above ~38°C.
  • Typical packaging sizes: 10 g, 25 g, 50 g, 100 g, and larger.
  • Applications include use as a carbon source and as a synthetic intermediate in assays and reactions.

Catalog No. 50-003-00778


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.