Learn More
Medchemexpress LLC Ethyl 4-isocyanatobenzoate-d4 | 1219802-91-1 | 98.0% | 195.21 g/mol | C10H5D4NO3 | 10 MG
SDP
Catalog No. 5000281858
Encompass_Preferred
Ethyl 4-isocyanatobenzoate-d4 is a deuterium-labeled analog of ethyl 4-isocyanatobenzoate supplied for use as a stable isotope standard and as a synthetic building block in organic synthesis and analytical workflows. Supplied as a white to off-white solid with high isotopic enrichment and analytical purity.
  • Deuterium-labeled standard for analytical quantitation.
  • High isotopic enrichment (99.7%).
  • High purity (98.0% by GC).
  • Stable solid form suitable for long-term storage.
  • Recommended storage: powder at -20°C; in solvent at -80°C for extended periods.
  • Handle with appropriate respiratory and skin protection due to irritation hazards.
Catalog No. 50-002-81858 Supplier Medchemexpress LLC Supplier No. HYW275290S10MG
Ethyl 4-isocyanatobenzoate-d4 is a deuterium-labeled analog of ethyl 4-isocyanatobenzoate supplied for use as a stable isotope standard and as a synthetic building block in organic synthesis and analytical workflows. Supplied as a white to off-white solid with high isotopic enrichment and analytical purity.
  • Deuterium-labeled standard for analytical quantitation.
  • High isotopic enrichment (99.7%).
  • High purity (98.0% by GC).
  • Stable solid form suitable for long-term storage.
  • Recommended storage: powder at -20°C; in solvent at -80°C for extended periods.
  • Handle with appropriate respiratory and skin protection due to irritation hazards.