Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl (4-Methoxybenzoyl)acetate 97.0+%, TCI America™
SDP

Catalog No. M138025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M138025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M138025G Supplier TCI America Supplier No. M138025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2881-83-6
Molecular Formula C12H14O4
Molecular Weight (g/mol) 222.24
MDL Number MFCD00449636
InChI Key KRAHENMBSVAAHD-UHFFFAOYSA-N
Synonym p-Anisoylacetic Acid Ethyl Ester, Ethyl p-Anisoylacetate, (4-Methoxybenzoyl)acetic Acid Ethyl Ester, Ethyl 3-(4-Methoxyphenyl)-3-oxopropionate, 3-(4-Methoxyphenyl)-3-oxopropionic Acid Ethyl Ester
PubChem CID 76150
IUPAC Name ethyl 3-(4-methoxyphenyl)-3-oxopropanoate
SMILES CCOC(=O)CC(=O)C1=CC=C(C=C1)OC

Specifications

pH 4.9 (1.74g/L H2O 19°C)
Boiling Point 170°C
Quantity 25 g
Formula Weight 222.24
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl (4-Methoxybenzoyl)acetate
EINECSNumber (3)-1745&(3)-1763
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.