Learn More
Medchemexpress LLC Ethyl 4'-((methyl(3-(pyridin-4-yl)benzyl)carbamoyl)oxy)-[1,1'-biphenyl]-4-carboxylate | 947669-86-5 | MFCD28160857 | 99.7% | 466.53 | C29H26N2O4 | 5 MG
SDP
Catalog No. 5000263530
Encompass_Preferred
WWL113 is a selective, orally active small-molecule inhibitor of carboxylesterases Ces3 and Ces1f used in biochemical and in vivo research to probe lipid metabolism and metabolic syndrome. It has shown efficacy in mouse models, improving lipid and glucose parameters after oral dosing.
  • Selective Ces3 and Ces1f inhibitor with potent activity.
  • Demonstrated in vivo efficacy in metabolic disease models.
  • High purity suitable for research applications.
  • Soluble in DMSO and formulatable for in vivo dosing protocols.
  • Applicable to biochemical, cell-based, and animal studies.
Catalog No. 50-002-63530 Supplier Medchemexpress LLC Supplier No. HY1101485MG
WWL113 is a selective, orally active small-molecule inhibitor of carboxylesterases Ces3 and Ces1f used in biochemical and in vivo research to probe lipid metabolism and metabolic syndrome. It has shown efficacy in mouse models, improving lipid and glucose parameters after oral dosing.
  • Selective Ces3 and Ces1f inhibitor with potent activity.
  • Demonstrated in vivo efficacy in metabolic disease models.
  • High purity suitable for research applications.
  • Soluble in DMSO and formulatable for in vivo dosing protocols.
  • Applicable to biochemical, cell-based, and animal studies.