Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethyl 4-methyloctanoate | 56196-53-3 | MFCD02262189 | 98.1% | 186.29 | C11H22O2 | 10 MG
SDP

Catalog No. 5000283366
Encompass_Preferred

Ethyl 4-methyloctanoate is an insect pheromone used as a research reagent in chemical ecology, behavioral assays, and analytical studies. It is provided as a high-purity ethyl ester suitable as a reference standard or synthesis intermediate.

  • High purity: 98.1%
  • Molecular weight: 186.29 g/mol
  • Chemical formula: C11H22O2
  • Available in small package sizes (5-200 mg)
  • Suitable for analytical and behavioral studies
  • Intended for laboratory research use only

Catalog No. 50-002-83366 Supplier Medchemexpress LLC Supplier No. HYW13129610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethyl 4-methyloctanoate is an insect pheromone used as a research reagent in chemical ecology, behavioral assays, and analytical studies. It is provided as a high-purity ethyl ester suitable as a reference standard or synthesis intermediate.

  • High purity: 98.1%
  • Molecular weight: 186.29 g/mol
  • Chemical formula: C11H22O2
  • Available in small package sizes (5-200 mg)
  • Suitable for analytical and behavioral studies
  • Intended for laboratory research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.