Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 4-Nitrophenylacetate 98.0+%, TCI America™
SDP

Catalog No. N022825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N022825G 25 g
N02285G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N022825G Supplier TCI America Supplier No. N022825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5445-26-1
Molecular Formula C10H11NO4
Molecular Weight (g/mol) 209.20
MDL Number MFCD00017046
InChI Key DWDRNKYLWMKWTH-UHFFFAOYSA-N
Synonym ethyl 4-nitrophenylacetate, ethyl 2-4-nitrophenyl acetate, ethyl p-nitrophenylacetate, ethyl 4-nitrobenzeneacetate, ethyl 4-nitrophenyl acetate, 4-nitrophenylacetic acid ethyl ester, benzeneacetic acid, 4-nitro-, ethyl ester, ethyl-4-nitrophenylacetate, acetic acid, p-nitrophenyl-, ethyl ester, 4-nitro-phenyl-acetic acid ethyl ester
PubChem CID 79517
IUPAC Name ethyl 2-(4-nitrophenyl)acetate
SMILES CCOC(=O)CC1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 64°C
Color White-Yellow
Boiling Point 196°C
Quantity 25 g
Formula Weight 209.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 4-Nitrophenylacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.