Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 4-(trifluoromethyl)benzoate, 99%

Catalog No. AAL1113903
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1113903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1113903 Supplier Thermo Scientific Chemicals Supplier No. L1113903
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 583-02-8 | C10H9F3O2 | 218.175 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 583-02-8
Molecular Formula C10H9F3O2
Molecular Weight (g/mol) 218.175
MDL Number MFCD00013563
InChI Key ZQDADDSPMCHZPX-UHFFFAOYSA-N
Synonym ethyl 4-trifluoromethyl benzoate, 4-trifluoromethyl benzoic acid ethyl ester, 4-trifluoromethylbenzoic acid, ethyl ester, pubchem16913, acmc-1cs5c, ethyl-p-trifluoromethylbenzoate, ethyl-4-trifluoromethyl benzoate, 4-trifluoromethylbenzoic acid ethyl ester
PubChem CID 521827
IUPAC Name ethyl 4-(trifluoromethyl)benzoate
SMILES CCOC(=O)C1=CC=C(C=C1)C(F)(F)F

Specifications

Boiling Point 78°C to 80°C (5 mmHg)
Refractive Index 1.447
Quantity 1 g
Beilstein 1968247
Formula Weight 218.18
Percent Purity 99%
Chemical Name or Material Ethyl 4-(trifluoromethyl)benzoate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA Yes

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.