Learn More
Accela Chembio Inc Ethyl (4-trifluoromethylbenzoyl)acetate | 1g | 106263-53-0 | MFCD03424818 | 97+% | D: 1.27 | Shelf Life: 1080 Days | Store In Freezer(-20) Upon Receipt
Supplier:  Accela Chembio Inc SY0039131G
Encompass_Preferred
Ethyl (4-trifluoromethylbenzoyl)acetate | 1g | 106263-53-0 | MFCD03424818 | 97+% | D: 1.27 | Shelf Life: 1080 Days | Store In Freezer(-20) Upon Receipt
Catalog No. 50-250-308