Learn More
Medchemexpress LLC Ethyl (4bS,8R,8aS,14bR)-7-(cyclopropylmethyl)-...-12-carboxylate | 892039-23-5 | 99.4% | C29H30N2O5 | 10MG
SDP
Catalog No. 5000261939
Encompass_Preferred
TAN-452 is an orally active, selective peripherally acting δ-opioid receptor (DOR) antagonist derived from naltrindole. It displays high potency at DOR (Ki 0.47 nM; Kb 0.21 nM), shows low brain penetrability, and can attenuate morphine-induced side effects without significantly reducing analgesia. The compound also exhibits antagonist activity at μ- and κ-opioid receptors at higher concentrations.
  • Selective peripheral DOR antagonist with subnanomolar potency.
  • Low brain penetrability reduces central nervous system exposure.
  • Attenuates morphine-induced side effects while preserving analgesia.
  • Also antagonizes μ- and κ-opioid receptors at higher concentrations.
  • White to off-white solid supplied with high reported purity.
Catalog No. 50-002-61939 Supplier Medchemexpress LLC Supplier No. HY13620810MG
TAN-452 is an orally active, selective peripherally acting δ-opioid receptor (DOR) antagonist derived from naltrindole. It displays high potency at DOR (Ki 0.47 nM; Kb 0.21 nM), shows low brain penetrability, and can attenuate morphine-induced side effects without significantly reducing analgesia. The compound also exhibits antagonist activity at μ- and κ-opioid receptors at higher concentrations.
  • Selective peripheral DOR antagonist with subnanomolar potency.
  • Low brain penetrability reduces central nervous system exposure.
  • Attenuates morphine-induced side effects while preserving analgesia.
  • Also antagonizes μ- and κ-opioid receptors at higher concentrations.
  • White to off-white solid supplied with high reported purity.