Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 5-amino-3-methylisothiazole-4-carboxylate, 97%, Thermo Scientific™

Catalog No. KM04205DA
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
KM04205DA 1 g
KM04205DE 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. KM04205DA Supplier Thermo Scientific Chemicals Supplier No. KM04205DA
Add to Cart

Chemical Identifiers

CAS 34859-65-9
Molecular Formula C7H10N2O2S
Molecular Weight (g/mol) 186.229
InChI Key ZDGBWOZFPKNORL-UHFFFAOYSA-N
PubChem CID 2820511
IUPAC Name ethyl 5-amino-3-methyl-1,2-thiazole-4-carboxylate
SMILES CCOC(=O)C1=C(SN=C1C)N

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym ethyl 5-amino-3-methylisothiazole-4-carboxylate, ethyl5-amino-3-methylisothiazole-4-carboxylate, ethyl 5-amino-3-methyl-isothiazole 4-carboxylate, maybridge1_004847, 5-amino-3-methyl-isothiazole-4-carboxylate ethyl ester, 5-amino-4-ethoxycarbonyl-3-methylisothiazole, 4-isothiazolecarboxylic acid, 5-amino-3-methyl-, ethyl ester, 4-isothiazolecarboxylicacid, 5-amino-3-methyl-, ethyl ester
Formula Weight 186.23
Percent Purity 97%
Chemical Name or Material Ethyl 5-amino-3-methylisothiazole-4-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.