Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethyl 5-aminobenzofuran-2-carboxylate | 174775-48-5 | 99.8% | C11H11NO3 | 10 G
SDP

Catalog No. 5000434491
Encompass_Preferred

Ethyl 5-aminobenzofuran-2-carboxylate, also known as Vilazodone Intermediate 5, is an off-white to brown solid used as a drug intermediate for the synthesis of various active compounds.

  • Purity of 99.8%
  • Appears as an off-white to brown solid
  • Functions as a drug intermediate
  • Store at 4°C, protected from light
  • For storage in solvent: -80°C for 6 months; -20°C for 1 month (protect from light)

Catalog No. 50-004-34491 Supplier Medchemexpress LLC Supplier No. HY7701610G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethyl 5-aminobenzofuran-2-carboxylate, also known as Vilazodone Intermediate 5, is an off-white to brown solid used as a drug intermediate for the synthesis of various active compounds.

  • Purity of 99.8%
  • Appears as an off-white to brown solid
  • Functions as a drug intermediate
  • Store at 4°C, protected from light
  • For storage in solvent: -80°C for 6 months; -20°C for 1 month (protect from light)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.