Learn More
Medchemexpress LLC Ethyl 5-aminobenzofuran-2-carboxylate | 174775-48-5 | 99.8% | C11H11NO3 | 25 G
SDP
Catalog No. 5000357632
Encompass_Preferred
Ethyl 5-aminobenzofuran-2-carboxylate (Vilazodone Intermediate 5) is a drug intermediate used for the synthesis of various active compounds. It is categorized as a drug intermediate and is intended for research use only, not for medical applications.
  • Drug intermediate for synthesis of active compounds
  • For research use only
Catalog No. 50-003-57632 Supplier Medchemexpress LLC Supplier No. 25GHY77016
Ethyl 5-aminobenzofuran-2-carboxylate (Vilazodone Intermediate 5) is a drug intermediate used for the synthesis of various active compounds. It is categorized as a drug intermediate and is intended for research use only, not for medical applications.
  • Drug intermediate for synthesis of active compounds
  • For research use only