Learn More
Medchemexpress LLC Ethyl 5-aminobenzofuran-2-carboxylate | 174775-48-5 | MFCD08275091 | >98.0% | C11H11NO3 | 5 G
SDP
Catalog No. 5000434497
Encompass_Preferred
Ethyl 5-aminobenzofuran-2-carboxylate serves as a synthetic intermediate for producing indolebutylpiperazines, which are dual 5-HT1A receptor agonists and serotonin reuptake inhibitors. It also finds use as a potential radiolabelled analog for melanoma imaging and targeted radiotherapy.
  • Utilized as a synthetic intermediate
  • Used in the creation of indolebutylpiperazines
  • Acts as a dual 5-HT1A receptor agonist
  • Functions as a serotonin reuptake inhibitor
  • Employed as a potential radiolabelled analog
  • Applied for melanoma imaging
  • Used for targeted radiotherapy
Catalog No. 50-004-34497 Supplier Medchemexpress LLC Supplier No. HY770165G
Ethyl 5-aminobenzofuran-2-carboxylate serves as a synthetic intermediate for producing indolebutylpiperazines, which are dual 5-HT1A receptor agonists and serotonin reuptake inhibitors. It also finds use as a potential radiolabelled analog for melanoma imaging and targeted radiotherapy.
  • Utilized as a synthetic intermediate
  • Used in the creation of indolebutylpiperazines
  • Acts as a dual 5-HT1A receptor agonist
  • Functions as a serotonin reuptake inhibitor
  • Employed as a potential radiolabelled analog
  • Applied for melanoma imaging
  • Used for targeted radiotherapy