Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 5-Bromo-2-chlorobenzoate 98.0+%, TCI America™
SDP

Catalog No. B22555G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B22555G 5 g
B225525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B22555G Supplier TCI America Supplier No. B22555G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 76008-73-6
Molecular Formula C9H8BrClO2
Molecular Weight (g/mol) 263.515
MDL Number MFCD00013635
InChI Key AMGWDYLEMSMUIO-UHFFFAOYSA-N
Synonym 5-bromo-2-chlorobenzoic acid ethyl ester, ethyl5-bromo-2-chlorobenzoate, benzoic acid, 5-bromo-2-chloro-, ethyl ester, pubchem3702, acmc-1bb9d, ethyl-5-bromo-2-chlorobenzoate, ethyl 5-bromo-2-chloro-benzoate, a-ethyl 5-bromo-2-chlorobenzoate, attercop-chm at115127, ethyl 5-bromo2-chlorobenzaldehyde
PubChem CID 144749
IUPAC Name ethyl 5-bromo-2-chlorobenzoate
SMILES CCOC(=O)C1=C(C=CC(=C1)Br)Cl

Specifications

Color Yellow
Boiling Point 288°C
Quantity 5 g
Formula Weight 263.52
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 5-Bromo-2-chlorobenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.