Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 5-Chloromethyl-2-furancarboxylate 97.0+%, TCI America™
SDP

Catalog No. C14945G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C14945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C14945G Supplier TCI America Supplier No. C14945G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2528-00-9
Molecular Formula C8H9ClO3
Molecular Weight (g/mol) 188.607
MDL Number MFCD00173837
InChI Key JBACYJRMCXLIQU-UHFFFAOYSA-N
Synonym ethyl 5-chloromethyl-2-furoate, ethyl 5-chloromethyl furan-2-carboxylate, ethyl 5-chloromethyl-2-furancarboxylate, 5-chloromethyl-2-furancarboxylic acid ethyl ester, 2-furancarboxylic acid, 5-chloromethyl-, ethyl ester, buttpark 9650-29, acmc-1cej4, ethyl 2-chloromethyl-5-furoate
PubChem CID 263166
IUPAC Name ethyl 5-(chloromethyl)furan-2-carboxylate
SMILES CCOC(=O)C1=CC=C(O1)CCl

Specifications

Boiling Point 140°C
Quantity 5 g
Formula Weight 188.61
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 5-Chloromethyl-2-furancarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.