Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 5-formyl-2,4-dimethylpyrrole-3-carboxylate, 95%, Thermo Scientific™

Catalog No. AAH6375103
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6375103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6375103 Supplier Thermo Scientific Chemicals Supplier No. H6375103

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2199-59-9
Molecular Formula C10H13NO3
Molecular Weight (g/mol) 195.22
MDL Number MFCD00030352
InChI Key GDISALBEIGGPER-UHFFFAOYSA-N
Synonym 5-formyl-2,4-dimethyl-1h-pyrrole-3-carboxylic acid ethyl ester, 2,4-dimethyl-3-ethoxycarbonyl-5-formylpyrrole, ethyl 2,4-dimethyl-5-formylpyrrole-3-carboxylate, ethyl 5-formyl-2,4-dimethylpyrrole-3-carboxylate, 1h-pyrrole-3-carboxylic acid, 5-formyl-2,4-dimethyl-, ethyl ester, ethyl 3,5-dimethyl-2-formylpyrrole-4-carboxylate, 2,4-dimethyl-3-carbethoxy pyrrole-5-carboxaldehyde, 3,5-dimethyl-4-ethoxycarbonyl-1h-pyrrole-2-carboxaldehyde, 5-formyl-2,4-dimethyl-3-pyrrolecarboxylic acid ethyl ester, pubchem9415
PubChem CID 137485
IUPAC Name ethyl 5-formyl-2,4-dimethyl-1H-pyrrole-3-carboxylate
SMILES CCOC(=O)C1=C(C)NC(C=O)=C1C

Specifications

Quantity 1 g
Formula Weight 195.22
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Ethyl 5-formyl-2,4-dimethylpyrrole-3-carboxylate
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.