Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 5-hydroxyindole-2-carboxylate, 98+%

Catalog No. AAL0077406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0077406 5 g
AAL0077403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0077406 Supplier Thermo Scientific Chemicals Supplier No. L0077406
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 24985-85-1 | C11H11NO3 | 205.213 g/mol

Ethyl 5-hydroxyindole-2-carboxylate is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 5-hydroxyindole-2-carboxylate is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

Solubility
Insoluble in water.

Notes
Store between 2-8 C. Store in cool, dryplace in a well ventilated area. Store in a tightly sealed container. Protect from moisture, heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24985-85-1
Molecular Formula C11H11NO3
Molecular Weight (g/mol) 205.213
MDL Number MFCD00015458
InChI Key WANAXLMRGYGCPC-UHFFFAOYSA-N
Synonym ethyl 5-hydroxyindole-2-carboxylate, 5-hydroxy-1h-indole-2-carboxylic acid ethyl ester, 1h-indole-2-carboxylic acid, 5-hydroxy-, ethyl ester, 5-hydroxyindole-2-carboxylic acid ethyl ester, ethyl-5-hydroxy-1h-indol-2-carboxylat, pubchem15850, acmc-1cs3a, 5-hydroxy-1h-indole-2-carboxylicacidethylester, 2-carbethoxy-5-hydroxy-indole, ethyl 5-hydroxy-1h-2-indolecarboxylate
PubChem CID 90677
IUPAC Name ethyl 5-hydroxy-1H-indole-2-carboxylate
SMILES CCOC(=O)C1=CC2=C(N1)C=CC(=C2)O

Specifications

Melting Point 150°C to 151°C
Quantity 5 g
Beilstein 476925
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 205.21
Percent Purity ≥98%
Chemical Name or Material Ethyl 5-hydroxyindole-2-carboxylate
EINECSNumber 246-554-9
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.