Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sigma Aldrich Ethyl 5-methyl-4-oxo-3,4-dihydrothieno[2,3-d]pyrimidine-6-carboxylate
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- Formaldehyde diethyl acetal (fdea, diethoxymethane, dem) is a diethyl acetal
- It has been synthesized from ethanol and aqueous formaldehyde
- It is a low boiling solvent useful for sodium hydride reactions, organolithium chemistry, copper-catalyzed conjugate additions and phase-transfer reactions
- It is a potential alternate for tetrahydrofuran, dichloromethane, glyme and methylal
- Dem is an ethoxymethylating agent, a formaldehyde equivalent and a carbonylation substrate
- Its condensation with 2-propylresorcinol to form resorcin[n]arenes (n=4-7) has been investigated
- The IR and raman spectra and conformations of dem have been studied
- NMR spectroscopy has been used to study the aggregation behavior of organolithium compounds in diethoxymethane
- Unimolecular metastable decomposition of dem upon electron impact has been studied
- Solubility of non-polar gases in dem has been evaluated.formaldehyde diethyl acetal may be used as a solvent in the esterification of carboxylic acids and in the o-alkylation of a variety of phenols
Specifications
Specifications
| CAS | 17417-67-3 |
| Molecular Formula | C10H10N2O3S |
| Linear Formula | C10H10N2O3S |
| MDL Number | MFCD00168240 |
| Quantity | 500 mL |
| Molecular Weight (g/mol) | 238.26 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?