Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethyl 6-(4-aminophenyl)-1-(4-methoxyphenyl)-7-oxo-4,5-dihydropyrazolo[3,4-c]pyridine-3-carbo | 503615-07-4 | 98.6% | C22H22N4O4 | 25 G
SDP

Catalog No. 5000357622
Encompass_Preferred

Ethyl 6-(4-aminophenyl)-1-(4-methoxyphenyl)-7-oxo-4,5-dihydropyrazolo[3,4-c]pyridine-3-carboxylate is a chemical compound primarily intended for research use. It presents as a light green to green solid, with a molecular formula of C22H22N4O4 and a molecular weight of 406.43. This product demonstrates a high purity of 98.61% as determined by LCMS.

  • Appearance: Light green to green solid
  • Purity (LCMS): 98.61%
  • Molecular formula: C22H22N4O4
  • Molecular weight: 406.43
  • Storage condition (solid): 4°C, protect from light
  • Storage condition (in solvent): -80°C for 6 months; -20°C for 1 month, protect from light

Catalog No. 50-003-57622 Supplier Medchemexpress LLC Supplier No. 25GHY75678
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethyl 6-(4-aminophenyl)-1-(4-methoxyphenyl)-7-oxo-4,5-dihydropyrazolo[3,4-c]pyridine-3-carboxylate is a chemical compound primarily intended for research use. It presents as a light green to green solid, with a molecular formula of C22H22N4O4 and a molecular weight of 406.43. This product demonstrates a high purity of 98.61% as determined by LCMS.

  • Appearance: Light green to green solid
  • Purity (LCMS): 98.61%
  • Molecular formula: C22H22N4O4
  • Molecular weight: 406.43
  • Storage condition (solid): 4°C, protect from light
  • Storage condition (in solvent): -80°C for 6 months; -20°C for 1 month, protect from light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.