Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethyl 6-(4-aminophenyl)-1-(4-methoxyphenyl)-7-oxo-4,5-dihydropyrazolo[3,4-c]pyridine-3-carbox | 503615-07-4 | MFCD18251628 | 98.6% | C22H22N4O4 | 10 G
SDP

Catalog No. 5000433612
Encompass_Preferred

This product is Ethyl 6-(4-aminophenyl)-1-(4-methoxyphenyl)-7-oxo-4,5-dihydropyrazolo[3,4-c]pyridine-3-carboxylate, a chemical compound that appears as a light green to green solid. It has been tested and complies with given specifications, showing a purity of 98.6% by LCMS. This product is for research use only and has not been fully validated for medical applications.

  • Appears as a light green to green solid
  • Purity of 98.6% confirmed by LCMS
  • Tested and complies with specifications
  • Store at 4°C, protected from light
  • Store in solvent at -80°C for up to 6 months or -20°C for up to 1 month, protected from light

Catalog No. 50-004-33612 Supplier Medchemexpress LLC Supplier No. HY7567810G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product is Ethyl 6-(4-aminophenyl)-1-(4-methoxyphenyl)-7-oxo-4,5-dihydropyrazolo[3,4-c]pyridine-3-carboxylate, a chemical compound that appears as a light green to green solid. It has been tested and complies with given specifications, showing a purity of 98.6% by LCMS. This product is for research use only and has not been fully validated for medical applications.

  • Appears as a light green to green solid
  • Purity of 98.6% confirmed by LCMS
  • Tested and complies with specifications
  • Store at 4°C, protected from light
  • Store in solvent at -80°C for up to 6 months or -20°C for up to 1 month, protected from light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.