Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethyl 6-(4-aminophenyl)-1-(4-methoxyphenyl)-7-oxo-4,5-dihydropyrazolo[3,4-c]pyridine | 503615-07-4 | 98.6% | C22H22N4O4 | 5 G
SDP

Catalog No. 5000433776
Encompass_Preferred

Ethyl 6-(4-aminophenyl)-1-(4-methoxyphenyl)-7-oxo-4,5-dihydropyrazolo[3,4-c]pyridine-3-carboxylate is a drug intermediate for the synthesis of various active compounds.

  • Solid, light green to green appearance.
  • Purity of 98.61% as determined by LCMS.
  • Store at 4°C, protected from light.
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light.

Catalog No. 50-004-33776 Supplier Medchemexpress LLC Supplier No. HY756785G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethyl 6-(4-aminophenyl)-1-(4-methoxyphenyl)-7-oxo-4,5-dihydropyrazolo[3,4-c]pyridine-3-carboxylate is a drug intermediate for the synthesis of various active compounds.

  • Solid, light green to green appearance.
  • Purity of 98.61% as determined by LCMS.
  • Store at 4°C, protected from light.
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.