Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 6-bromo-4-chloroquinoline-3-carboxylate, 97%, Thermo Scientific Chemicals

Catalog No. AAH5930103
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5930103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5930103 Supplier Thermo Scientific Chemicals Supplier No. H5930103
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 206257-39-8 | Molecular Formula: C12H9BrClNO2 | Molecular Weight: 314.56 g/mol

Ethyl 6-bromo-4-chloroquinoline-3-carboxylate is used as pharmaceutical intermediates

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 6-bromo-4-chloroquinoline-3-carboxylate is used as pharmaceutical intermediates

Solubility
Immiscible in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 206257-39-8
Molecular Formula C12H9BrClNO2
Molecular Weight (g/mol) 314.56
MDL Number MFCD00173367
InChI Key YGMLKBFPHVLHGT-UHFFFAOYSA-N
Synonym ethyl 6-bromo-4-chloro-3-quinolinecarboxylate, 4-chloro-6-bromoquinoline-3-carboxylic acid ethyl ester, 3-quinolinecarboxylic acid, 6-bromo-4-chloro-, ethyl ester, 6-bromo-4-chloroquinoline-3-carboxylic acid ethyl ester, ethyl 6-bromo-4-chloro-quinoline-3-carboxylate, ethylbromochloroquinolinecarboxylate, ethyl-6-bromo-4-chloro-quinoline-3-carboxylate, 6-bromo-4-chloro-3-quinolinecarboxylic acid ethyl ester, 6-bromo-4-chloro-quinoline-3-carboxylic acid ethyl ester
PubChem CID 2764138
IUPAC Name ethyl 6-bromo-4-chloroquinoline-3-carboxylate
SMILES CCOC(=O)C1=CN=C2C=CC(Br)=CC2=C1Cl

Specifications

Quantity 1 g
Solubility Information Immiscible in water.
Formula Weight 314.57
Percent Purity 97%
Chemical Name or Material Ethyl 6-bromo-4-chloroquinoline-3-carboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.