Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 6-bromohexanoate, 98%

Catalog No. AC305921000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC305921000 100 g Glass bottle
AC305920250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC305921000 Supplier Thermo Scientific Chemicals Supplier No. 305921000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 25542-62-5 | C8H15BrO2 | 223.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25542-62-5
Molecular Formula C8H15BrO2
Molecular Weight (g/mol) 223.11
MDL Number MFCD00000270
InChI Key DXBULVYHTICWKT-UHFFFAOYSA-N
Synonym 6-bromohexanoic acid ethyl ester, ethyl 6-bromocaproate, ethyl 6-bromocapronate, 6-bromocaproic acid ethyl ester, ethyl omega-bromocaproate, hexanoic acid, 6-bromo-, ethyl ester, ethyl .omega.-bromocaproate, 6-bromo-hexanoic acid ethyl ester, ethyl 6bromohexanoate, pubchem3953
PubChem CID 117544
IUPAC Name ethyl 6-bromohexanoate
SMILES CCOC(=O)CCCCCBr

Specifications

Color White to Beige
Density 1.2540g/mL
Boiling Point 128°C to 130°C (16.0 mmHg)
Flash Point 58,3°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula Br(CH2)5CO2C2H5
Refractive Index 1.4580 to 1.46
Quantity 100 g
Specific Gravity 1.254
Formula Weight 223.11
Percent Purity 98%
Physical Form Powder or Granular Powder
Chemical Name or Material Ethyl 6-bromohexanoate, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 247-085-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.