Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethyl (6Z,9Z,12Z,15Z,18Z)-6,9,12,15,18-heneicosapentaenoate | 131775-86-5 | 94.0% | 344.5 g/mol | C23H36O2 | 10MG
SDP

Catalog No. 5000237772
Encompass_Preferred

Ethyl (6Z,9Z,12Z,15Z,18Z)-6,9,12,15,18-heneicosapentaenoate is the ethyl ester derivative of heneicosapentaenoic acid, an omega-3 polyunsaturated fatty acid. It is supplied as a research chemical and reference standard for biochemical and lipid metabolism studies. Purity is vendor-dependent and should be confirmed with the certificate of analysis.

  • Molecular formula C23H36O2 and molecular weight 344.5 g/mol.
  • Identified by CAS 131775-86-5 and InChIKey RKAKAVQIMCZXPE-AAQCHOMXSA-N.
  • Used as a research chemical and reference standard in lipid and metabolism studies.
  • Available in small research pack sizes suitable for analytical and reference use.
  • Soluble in common organic solvents such as chloroform and ethyl acetate.
  • Purity varies by supplier; confirm certificate of analysis prior to use.

Catalog No. 50-002-37772 Supplier Medchemexpress LLC Supplier No. HY12411810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethyl (6Z,9Z,12Z,15Z,18Z)-6,9,12,15,18-heneicosapentaenoate is the ethyl ester derivative of heneicosapentaenoic acid, an omega-3 polyunsaturated fatty acid. It is supplied as a research chemical and reference standard for biochemical and lipid metabolism studies. Purity is vendor-dependent and should be confirmed with the certificate of analysis.

  • Molecular formula C23H36O2 and molecular weight 344.5 g/mol.
  • Identified by CAS 131775-86-5 and InChIKey RKAKAVQIMCZXPE-AAQCHOMXSA-N.
  • Used as a research chemical and reference standard in lipid and metabolism studies.
  • Available in small research pack sizes suitable for analytical and reference use.
  • Soluble in common organic solvents such as chloroform and ethyl acetate.
  • Purity varies by supplier; confirm certificate of analysis prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.