Learn More
Medchemexpress LLC Ethyl 7-bromoheptanoate | 29823-18-5 | 237.13 | 10 G
SDP
Catalog No. 5000447879
Encompass_Preferred
Ethyl 7-bromoheptanoate is a key intermediate and alkylating agent used in the synthesis of other active compounds, including prostaglandins and their analogues. It can also react with quinazoline derivatives containing hydroxyl groups to introduce ester group-containing side chains.
  • Liquid appearance (density: 1.223±0.06 g/cm3)
  • Colorless to light yellow
Catalog No. 50-004-47879 Supplier Medchemexpress LLC Supplier No. HYW00824510G
Ethyl 7-bromoheptanoate is a key intermediate and alkylating agent used in the synthesis of other active compounds, including prostaglandins and their analogues. It can also react with quinazoline derivatives containing hydroxyl groups to introduce ester group-containing side chains.
  • Liquid appearance (density: 1.223±0.06 g/cm3)
  • Colorless to light yellow