Learn More
Medchemexpress LLC Ethyl 7-bromoheptanoate | 29823-18-5 | 237.13 | 25 G
SDP
Catalog No. 5000448068
Encompass_Preferred
Ethyl 7-bromoheptanoate is a key intermediate and alkylating agent used in the synthesis of other active compounds. It can be utilized in the synthesis of prostaglandins and their analogues, and can react with quinazoline derivatives containing hydroxyl groups to introduce ester group-containing side chains.
  • Key intermediate
  • Alkylating agent
  • Used in the synthesis of prostaglandins and their analogues
  • Can introduce ester group-containing side chains to quinazoline derivatives
Catalog No. 50-004-48068 Supplier Medchemexpress LLC Supplier No. 25GHYW008245
Ethyl 7-bromoheptanoate is a key intermediate and alkylating agent used in the synthesis of other active compounds. It can be utilized in the synthesis of prostaglandins and their analogues, and can react with quinazoline derivatives containing hydroxyl groups to introduce ester group-containing side chains.
  • Key intermediate
  • Alkylating agent
  • Used in the synthesis of prostaglandins and their analogues
  • Can introduce ester group-containing side chains to quinazoline derivatives