Learn More
Apexbio Technology LLC Ethyl 7-(diethylamino)-2-oxo-2H-chromene-3-carboxylate 28705-46-6 1g
SDP
Catalog No. 5000095953
Encompass_Preferred
Ethyl 7-(diethylamino)-2-oxo-2H-chromene-3-carboxylate (CAS 28705-46-6) is a coumarin-based derivative that functions as a selective inhibitor of human monoamine oxidase B (hMAO-B) By inhibiting hMAO-B activity this compound reduces the enzymatic degradation of dopamine resulting in elevated dopaminergic levels in neural tissues Due to its selectivity it serves as a valuable tool in studies investigating dopamine metabolism and the pathophysiology of neurodegenerative disorders such as Parkinson s disease and depressive syndromes Its use supports research into MAO-B-targeted therapeutic strategies and neuroprotection mechanisms
Catalog No. 50-000-95953 Supplier Apexbio Technology LLC Supplier No. A99761000
Ethyl 7-(diethylamino)-2-oxo-2H-chromene-3-carboxylate (CAS 28705-46-6) is a coumarin-based derivative that functions as a selective inhibitor of human monoamine oxidase B (hMAO-B) By inhibiting hMAO-B activity this compound reduces the enzymatic degradation of dopamine resulting in elevated dopaminergic levels in neural tissues Due to its selectivity it serves as a valuable tool in studies investigating dopamine metabolism and the pathophysiology of neurodegenerative disorders such as Parkinson s disease and depressive syndromes Its use supports research into MAO-B-targeted therapeutic strategies and neuroprotection mechanisms